Order Entry
ContactUsLinkComponent
Anti-CD20 Rabbit Recombinant Antibody [clone: IGEL/7015R]
Anti-CD20 Rabbit Recombinant Antibody [clone: IGEL/7015R]
Catalog #: 76957-910
Supplier:  ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-CD20 IGEL7015R BSA 100UG
Anti-CD20 Rabbit Recombinant Antibody [clone: IGEL/7015R]
Catalog #: 76957-910
Supplier:  ANTIBODIES.COM LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

About this item

Recombinant rabbit monoclonal [IGEL/7015R] antibody to CD20 for WB with samples derived from Human and Canine.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1 to 2 µg/ml

Type: Primary
Antigen: CD20
Clonality: Recombinant
Clone: IGEL/7015R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Dog

Customers Who Bought This Also Bought