About this item
Mouse monoclonal [L26 + IGEL/773] antibody to CD20 for Flow Cytometry, IF and IHC-P with samples derived from Human, Baboon and Monkey.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 5 to 10 µl / million cells, IF: 1:50 to 1:100, IHC-P: 1:100 to 1:200
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: L26 + IGEL/773
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human;Baboon;Monkey
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76961-760
- 76951-756
- Antibody type:
- Primary
- Primary