Order Entry
ContactUsLinkComponent
Anti-CD20 Mouse Monoclonal Antibody [clone: 93-1B3]
Anti-CD20 Mouse Monoclonal Antibody [clone: 93-1B3]
Catalog #: 76951-570
Supplier:  ANTIBODIES.COM LLC
New Product
Anti-CD20 Mouse Monoclonal Antibody [clone: 93-1B3]
Catalog #: 76951-570
Supplier:  ANTIBODIES.COM LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Cat. no.:
    76951-570

Specifications

About this item

Mouse monoclonal [93-1B3] antibody to CD20 for Functional Studies, Flow Cytometry and IF with samples derived from Human.

  • Validated Applications: Functional Studies, Flow Cytometry, IF
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 93-1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Customers Who Bought This Also Bought