About this item
Recombinant rabbit monoclonal [DM193] antibody to CD45 for ELISA and Flow Cytometry with samples derived from Human.
- Validated Applications: ELISA, Flow Cytometry
- Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100
Type: Primary
Antigen: CD45
Clonality: Recombinant
Clone: DM193
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77186-702
- 77186-704
- Antigen name:
- B220
- B220
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Clone:
- DM193
- DM193
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# P08575
- UniprotID# P08575
- Isotype:
- IgG
- IgG