To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Amine oxidase (flavin containing) domain 2
- Clonality:Monoclonal
- Clone:PCRP-KDM1A-1A10
- Gene ID:UniprotID# O60341
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:KDM1 / LSD1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:EC1|BHC110|BRAF35-HDAC complex protein BHC110|BRAF35 HDAC complex protein BHC110|CPRF|AOF2|Amine oxidase, flavin containing, 2|FAD binding protein BRAF35 HDAC complex, 110 kDa subunit|BRAF35/HDAC complex, 110-kD subunit
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:92.9 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 152-279) of human KDM1A protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-KDM1A-1A10] antibody to KDM1 / LSD1 for Flow Cytometry, WB and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: KDM1 / LSD1
Clonality: Monoclonal
Clone: PCRP-KDM1A-1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human