To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:binding peptide
- Clonality:Monoclonal
- Clone:HP/3832
- Gene ID:UniprotID# P00738
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:Haptoglobin
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Haptoglobin alpha chain|Haptoglobin beta chain|Haptoglobin alpha(2FS) beta|Haptoglobin alpha(1S) beta|Haptoglobin|haptoglobin alpha polypeptide|BP|haptoglobin alpha 1S beta|haptoglobin alpha 2FS beta
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:15 - 18 kDa ( chain)/45 kDa ( chain)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 100-250) of human HP protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [HP/3832] antibody to Haptoglobin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Haptoglobin
Clonality: Monoclonal
Clone: HP/3832
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human