Anti-Dopamine beta Hydroxylase Mouse Monoclonal Antibody [clone: DBH/7222]
Catalog # 77179-468
Supplier: ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:dbh
- Antigen symbol:Dopamine beta Hydroxylase
- Clonality:Monoclonal
- Clone:DBH/7222
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P09172
- Antigen synonyms:Dopamine beta-monooxygenase|Dopamine beta monooxygenase|DBM|Soluble dopamine beta-hydroxylase|OTTHUMP00000022501|Dopamine beta-hydroxylase (dopamine beta-monooxygenase)|DOPO_HUMAN|Dopamine beta Hydroxylase
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:78 kDa/84 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 200-400) of human DBH protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77179-468
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [DBH/7222] antibody to Dopamine beta Hydroxylase for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Dopamine beta Hydroxylase
Clonality: Monoclonal
Clone: DBH/7222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human