Anti-FAM129B Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to FAM129B for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: FAM129B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77231-680
- Antigen symbol:
- FAM129B
- Antigen name:
- bA356B19.6
- Antigen synonyms:
- hypothetical protein LOC64855|FLJ22151|FLJ13518|family with sequence similarity 129, member B|chromosome 9 open reading frame 88|DKFZP434H0820|C9orf88|FAM129B|FLJ22298
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q96TA1
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 447-746 of human FAM129B (NP_073744.2).
- Sequence:
- TFETLLHQELGKGPTKEELCKSIQRVLERVLKKYDYDSSSVRKRFFREALLQISIPFLLKKLAPTCKSELPRFQELIFEDFARFILVENTYEEVVLQTVMKDILQAVKEAAVQRKHNLYRDSMVMHNSDPNLHLLAEGAPIDWGEEYSNSGGGGSPSPSTPESATLSEKRRRAKQVVSVVQDEEVGLPFEASPESPPPASPDGVTEIRGLLAQGLRPESPPPAGPLLNGAPAGESPQPKAAPEASSPPASPLQHLLPGKAVDLGPPKPSDQETGEQVSSPSSHPALHTTTEDSAGVQTEF
- Form:
- Liquid
- Molecular weight:
- 95 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
Frequently Bought Together