To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Anterior pituitary glycoprotein hormones common subunit alpha
- Clonality:Monoclonal
- Clone:hCGa/7138
- Gene ID:UniprotID# P01215
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:HCG alpha
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Follicle-stimulating hormone alpha|Chorionic Gonadotrophin Alpha Subunit|CG alpha|Chorionic Gonadotrophin Alpha|Choriogonadotropin alpha chain|CG-alpha|cga|Chorionic gonadotrophin subunit alpha|chorionic gonadotropin, alpha polypeptide
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:13 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-116) of human hCG alpha protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-044
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [hCGa/7138] antibody to HCG alpha for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: HCG alpha
Clonality: Monoclonal
Clone: hCGa/7138
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human