Anti-ATMIN Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to ATMIN for WB and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:100
Type: Primary
Antigen: ATMIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76856-690
- Antigen symbol:
- ATMIN
- Antigen name:
- ATM Interacting protein
- Antigen synonyms:
- ATM/ATR substrate CHEK2 interacting zinc finger protein|ATM interactor|ATM/ATR substrate Chk2 interacting Zn2+ finger protein|ATM/ATR Substrate Chk2 Interacting Zn++ finger protein|ATM substrate CHK2-interacting zinc finger protein|ATMIN|ATM/ATR-substrate CHK2-interacting zinc finger protein|ATMIN_HUMAN|ASCIZ
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# O43313
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 160 - 360 of human ATMIN (NP_056066.2)
- Sequence:
- HAEKKHKCSKCSNSYGTEWDLKRHAEDCGKTFRCTCGCPYASRTALQSHIYRTGHEIPAEHRDPPSKKRKMENCAQNQKLSNKTIESLNNQPIPRPDTQELEASEIKLEPSFEDSCGSNTDKQTLTTPPRYPQKLLLPKPKVALVKLPVMQFSVMPVFVPTADSSAQPVVLGVDQGSATGAVHLMPLSVGTLILGLDSEAC
- Form:
- Liquid
- Molecular weight:
- 68 - 88 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Tested applications:
- ICC/IF
Specifications
Frequently Bought Together