To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fc epsilon RI
- Clonality:Monoclonal
- Clone:CRA1
- Gene ID:UniprotID# P12319
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:FCER1A
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:Amino acids 26 - 110
- Form:Liquid
- Antigen synonyms:alpha|Fc fragment of IgE, high affinity I, receptor for, alpha subunit|Fc fragment of IgE, high affinity I, receptor for|Fc IgE receptor, alpha polypeptide|alpha-chain|Fc fragment of IgE high affinity I receptor for alpha polypeptide|Fc IgE receptor alpha polypeptide|Fc IgE receptor, alpha chain|alpha chain
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol (filter-sterilized, without Sodium Azide and carrier free)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant extracellular portion of human FCER1A protein (corresponding to a.a. Met-26-197, where signal peptide is 1-25)
- Tested applications:IHC-P, IHC-Fr
- Purification:Affinity purification
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal (CRA1) antibody to FCER1A for WB, IP, FC, IHC-P and IHC-Fr with samples derived from Human.
- Validated Applications: WB, IP, FC, IHC-P, IHC-Fr
- Recommended Dilutions: WB: 1 µg/ml, FC: 1 to 5 µg/ml
Type: Primary
Antigen: FCER1A
Clonality: Monoclonal
Clone: CRA1
Conjugation: Unconjugated
Epitope: Amino acids 26-110.
Host: Mouse
Isotype: IgG2b
Reactivity: Human