To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:APY
- Clonality:Recombinant
- Gene ID:UniprotID# P11836
- Host:Recombinant
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CD20
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:100 µg
- Form:Lyophilized
- Antigen synonyms:B-lymphocyte surface antigen B1|B Lymphocyte Cell Surface Antigen B1|Bp35|ATOPY|CD 20|B-lymphocyte antigen CD20|B-lymphocyte cell-surface antigen B1|Bp 35|B lymphocyte antigen CD20
- Storage buffer:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage temperature:Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Concentration:Reconstitution dependent
- Shipping temperature:Shipped on blue ice at 4 °C
- Packaging:Plastic vial
Specifications
About this item
Recombinant humanized monoclonal [Obinutuzumab Biosimilar] antibody to CD20 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: CD20
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human