About this item
Recombinant chimeric monoclonal [DMC686] antibody to Placental Alkaline Phosphatase/PLAP for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: Placental Alkaline Phosphatase/PLAP
Clonality: Recombinant
Clone: DMC686
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77187-366
- 77187-368
- Antigen name:
- Placental Alkaline Phosphatase/PLAP
- Placental Alkaline Phosphatase/PLAP
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Clone:
- DMC686
- DMC686
- Host:
- Recombinant
- Recombinant
- Gene ID:
- UniprotID# P05187
- UniprotID# P05187
- Isotype:
- IgG1
- IgG1