To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:AWT1
- Clonality:Monoclonal
- Clone:WT1/857 + 6F-H2
- Gene ID:UniprotID# P19544
- Host:Mouse
- Isotype:Multiple
- Reactivity:Human,Rat
- Antigen symbol:Wilms Tumor Protein
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Wilms Tumor Protein|WAGR|FWT1|NPHS4|Wilms' tumor protein|Wilms Tumor|GUD|Wilms' tumor gene|Wilms tumor 1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:47 - 55 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Clone WT1/857: Recombinant full - length human WT1 protein. Clone 6F - H2: Recombinant fragment, corresponding to amino acids 1 - 181, of human WT1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [WT1/857 + 6F-H2] antibody to Wilms Tumor Protein for IHC-P with samples derived from Human and Rat.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Wilms Tumor Protein
Clonality: Monoclonal
Clone: WT1/857 + 6F-H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human;Rat