To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:bHLHa17
- Clonality:Monoclonal
- Clone:TAL1/2707
- Gene ID:UniprotID# P17542
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Antigen symbol:Tal1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Stem cell protein|SCL|T cell acute lymphocytic leukemia 1 protein|STEM CELL LEUKEMIA HEMATOPOIETIC TRANSCRIPTION FACTOR|T cell leukemia/lymphoma 5 protein|OTTHUMP00000009564|Class A basic helix-loop-helix protein 17|T cell acute lymphocytic leukemia 1|OTTHUMP00000009563
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:42 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human TAL1 protein
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [TAL1/2707] antibody to Tal1 for ELISA with samples derived from Human.
- Validated Applications: ELISA
Type: Primary
Antigen: Tal1
Clonality: Monoclonal
Clone: TAL1/2707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human