Order Entry
ContactUsLinkComponent
Anti-SOX2 Rabbit Recombinant Antibody [clone: SOX2/3169R]
Anti-SOX2 Rabbit Recombinant Antibody [clone: SOX2/3169R]
Catalog #: 76965-104
Supplier:  ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-SOX2 SOX23169R BSA 100UG
Anti-SOX2 Rabbit Recombinant Antibody [clone: SOX2/3169R]
Catalog #: 76965-104
Supplier:  ANTIBODIES.COM LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 µg

Specifications

About this item

Recombinant rabbit monoclonal [SOX2/3169R] antibody to SOX2 for ELISA, WB and IHC-P with samples derived from Human and Mouse.

  • Validated Applications: ELISA, WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: SOX2
Clonality: Recombinant
Clone: SOX2/3169R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse

Customers Who Bought This Also Bought