To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Choreoathetosis/spasticity episodic (paroxysmal choreoathetosis/spasticity)
- Clonality:Monoclonal
- Clone:GLUT1/2475
- Gene ID:UniprotID# P11166
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Antigen symbol:GLUT1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:EIG12|erythrocyte/brain|Glucose transporter 1|facilitated glucose transporter member 1|DYT18|DYT9|Erythrocyte/hepatoma glucose transporter|DYT17|CSE
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:55 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 203 - 305, of human GLUT1 protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [GLUT1/2475] antibody to GLUT1 for ELISA, Flow Cytometry, IF, WB and IHC-P with samples derived from Human.
- Validated Applications: ELISA, Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: ELISA: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: GLUT1
Clonality: Monoclonal
Clone: GLUT1/2475
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human