Anti-gamma Parvin Mouse Monoclonal Antibody [clone: 8C5.2]
Catalog # 76959-214
Supplier: ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen symbol:gamma Parvin
- Clonality:Monoclonal
- Clone:8C5.2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9HBI0
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:37 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:GST-tagged recombinant protein corresponding to human ga mMa Parvin
- Purification:Protein A/G chromatography
- Cat. no.:76959-214
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [8C5.2] antibody to gamma Parvin for IP and WB with samples derived from Human.
- Validated Applications: IP, WB
- Recommended Dilutions: IP: 1 to 2 µg / 500 µg protein lysate, WB: 1 to 2 µg/ml
Type: Primary
Antigen: gamma Parvin
Clonality: Monoclonal
Clone: 8C5.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human