To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CAD16_HUMAN
- Clonality:Monoclonal
- Clone:CDH16/2125
- Gene ID:UniprotID# O75309
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:Cadherin 16
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CDH16|Cadherin-16|CDH 16|Kidney specific cadherin|Cadherin 16 KSP cadherin|Cadherin 16|Kidney cadherin|Cadherin kidney|Kidney-specific cadherin
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:130 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 371 - 507, of human CDH16 protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [CDH16/2125] antibody to Cadherin 16 for ELISA, WB and IHC-P with samples derived from Human.
- Validated Applications: ELISA, WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cadherin 16
Clonality: Monoclonal
Clone: CDH16/2125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human