To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:APY
- Clonality:Monoclonal
- Clone:109-3C2
- Gene ID:UniprotID# P11836
- Host:Mouse
- Isotype:IgG3 kappa
- Reactivity:Human
- Antigen symbol:CD20
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:B-lymphocyte surface antigen B1|B Lymphocyte Cell Surface Antigen B1|Bp35|ATOPY|CD 20|B-lymphocyte antigen CD20|B-lymphocyte cell-surface antigen B1|Bp 35|B lymphocyte antigen CD20
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:33 - 37 kDa
- Storage temperature:Shipped at 4 ℃. Store at 4 ℃.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Stimulated human leukocytes
- Tested applications:Functional Studies
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [109-3C2] antibody to CD20 for Functional Studies and ELISA with samples derived from Human.
- Validated Applications: Functional Studies, ELISA
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 109-3C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human