To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ANCR
- Clonality:Monoclonal
- Clone:PCRP-UBE3A-1A2
- Gene ID:UniprotID# Q05086
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:UBE3A
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:E6AP|E6AP ubiquitin protein ligase|CTCL tumor antigen se37-2|E6 AP|Angelman syndrome|EPVE6AP|AS|CTCL tumor antigen se37 2|E6AP ubiquitin-protein ligase
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:100 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human UBE3A protein
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-UBE3A-1A2] antibody to UBE3A for Flow Cytometry and IF with samples derived from Human.
- Validated Applications: Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: UBE3A
Clonality: Monoclonal
Clone: PCRP-UBE3A-1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human