To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:B220
- Clonality:Monoclonal
- Clone:2B11 + PD7/26
- Gene ID:UniprotID# P08575
- Host:Mouse
- Isotype:Multiple
- Reactivity:Human,Dog
- Antigen symbol:CD45
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CD45R|CD45|GP180|L-CA|Leukocyte Common antigen|LCA|loc|CD45 antigen|CD 45
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:180 - 220 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Clone 2B11: Isolated neoplastic cells from T cell lymphoma. Clone PD7/26: Human peripheral blood lymphocytes maintained in T cell growth factor
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [2B11 + PD7/26] antibody to CD45 for Flow Cytometry, WB and IHC-P with samples derived from Human and Canine.
- Validated Applications: Flow Cytometry, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 2B11 + PD7/26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human;Dog