Order Entry
ContactUsLinkComponent
Anti-Tartrate Resistant Acid Phosphatase Rabbit Recombinant Antibody [clone: ACP5/2336R]
Anti-Tartrate Resistant Acid Phosphatase Rabbit Recombinant Antibody [clone: ACP5/2336R]
Catalog #: 76955-890
Supplier:  ANTIBODIES.COM LLC
New Product
ANTIBODY A249691-100 100UG
Anti-Tartrate Resistant Acid Phosphatase Rabbit Recombinant Antibody [clone: ACP5/2336R]
Catalog #: 76955-890
Supplier:  ANTIBODIES.COM LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 µg

Specifications

About this item

Recombinant rabbit monoclonal [ACP5/2336R] antibody to Tartrate Resistant Acid Phosphatase for IHC-P with samples derived from Human, Mouse and Rat.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Tartrate Resistant Acid Phosphatase
Clonality: Recombinant
Clone: ACP5/2336R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat

Customers Who Bought This Also Bought