To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ADA-binding protein
- Clonality:Monoclonal
- Clone:DPP4/910
- Gene ID:UniprotID# P27487
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Rat
- Antigen symbol:CD26
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:ADCP 2|CD26 antigen 3|ADCP2|Adenosine deaminase complexing protein 2|CD 26|ADABP|CD26 antigen|ADCP-2|CD26
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:110 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant human DPP4 protein
- Tested applications:IHC
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [DPP4/910] antibody to CD26 for Flow Cytometry, IF and IHC with samples derived from Human and Rat.
- Validated Applications: Flow Cytometry, IF, IHC
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: CD26
Clonality: Monoclonal
Clone: DPP4/910
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Rat