To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:APY
- Clonality:Monoclonal
- Clone:2H7
- Gene ID:UniprotID# P11836
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Non-Human Primates,Human
- Antigen symbol:CD20
- Conjugation:APC (Allophycocyanin)-Cy7®
- Flow cytometry:Yes
- Size:100 tests
- Form:Liquid
- Antigen synonyms:B-lymphocyte surface antigen B1|B Lymphocyte Cell Surface Antigen B1|Bp35|ATOPY|CD 20|B-lymphocyte antigen CD20|B-lymphocyte cell-surface antigen B1|Bp 35|B lymphocyte antigen CD20
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 ℃. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Human tonsillar B cells
- Purification:This antibody is conjugated with tandem dye APC-Cyanine 7 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [2H7] antibody to CD20 (APC-Cyanine 7) for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Non-Human Primates