Anti-GST Goat Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Goat polyclonal antibody to GST for WB with samples derived from Schistosoma japonicum.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Schistosoma japonicum
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76835-682
- 76833-954
- Antigen symbol:
- GST
- GST
- Antigen name:
- Glutathione S Transferase
- Glutathione S Transferase
- Antigen synonyms:
- Glutathione S-transferase class-mu 26 kDa isozyme|GST epsilon|GST class alpha 2|GST|GST class alpha 1|GST gamma|Glutathione S transferase A1|Glutathione S transferase A2|GSTA1
- Glutathione S-transferase class-mu 26 kDa isozyme|GST epsilon|GST class alpha 2|GST|GST class alpha 1|GST gamma|Glutathione S transferase A1|Glutathione S transferase A2|GSTA1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Schistosoma japonicum
- Schistosoma japonicum
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Immunogen:
- Recombinant Glutathione-S-Transferase, expressed in and purified from E. coli
- Recombinant Glutathione-S-Transferase, expressed in and purified from E. coli
- Form:
- Liquid
- Liquid
- Purification:
- Affinity purification
- Affinity purification
- Concentration:
- 6 mg/ml
- 6 mg/ml
- Storage buffer:
- Supplied in Phosphate Buffered Saline with 20% Glycerol and 0.05% Sodium Azide
- Supplied in Phosphate Buffered Saline with 20% Glycerol and 0.05% Sodium Azide
- Storage temperature:
- Shipped at 4 ℃. Upon delivery aliquot and store at 4 ℃. Avoid freeze/thaw cycles.
- Shipped at 4 ℃. Upon delivery aliquot and store at 4 ℃. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 ℃
- Shipped on blue ice at 4 ℃
Specifications
Frequently Bought Together