Anti-MED22 Mouse Monoclonal Antibody [Clone: PCRP-MED22-1E4]
76957-100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:MED22
- Antigen symbol:MED22
- Clonality:Monoclonal
- Clone:PCRP-MED22-1E4
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2c
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q15528
- Antigen synonyms:Mediator complex subunit 22|Surf-5|Surf 5|OTTHUMP00000022480|RGD1564893|OTTMUSP00000012503|MED22_HUMAN|Mediator of RNA polymerase II transcription subunit 22|MED24
- Storage buffer:Supplied in 10mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:22 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human MED22 protein.
- Purification:Protein A/G chromatography
- Cat. no.:76957-100
- Packaging:Plastic vial
Mouse monoclonal [PCRP-MED22-1E4] antibody to MED22 for ELISA, IF, IP, Flow Cytometry and WB with samples derived from Human.
- Validated Applications: ELISA, IF, IP, Flow Cytometry, WB
- Recommended Dilutions: IF: 0.5 to 2 µg/ml, IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 0.5 to 2 µg/million cells, WB: 1 to 2 µg/ml
Type: Primary
Antigen: MED22
Clonality: Monoclonal
Clone: PCRP-MED22-1E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c
Reactivity: Human