To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CCN1
- Clonality:Monoclonal
- Clone:E67
- Gene ID:UniprotID# P20248
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:African green monkey,Human,Bovine,Mink,Mouse
- Antigen symbol:Cyclin A2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Cyclin A2|CCNA|Ccna2|Cyclin-A|CCNA2_HUMAN|Cyclin-A2
- Storage buffer:Supplied in 10mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:54 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:full-length bovine Cyclin A protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [E67] antibody to Cyclin A2 for Flow Cytometry, IF and IHC-P with samples derived from Human, Mouse, Bovine, African Green Monkey and Mink.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cyclin A2
Clonality: Monoclonal
Clone: E67
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Bovine, African Green monkey, Mink