Order Entry
ContactUsLinkComponent
Anti-MMP16 Rabbit Polyclonal Antibody
Anti-MMP16 Rabbit Polyclonal Antibody
Catalog #: 77050-996
Supplier:  ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-MMP-16 100UG
Anti-MMP16 Rabbit Polyclonal Antibody
Catalog #: 77050-996
Supplier:  ANTIBODIES.COM LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C8orf57
  • Clonality:
    Polyclonal
  • Gene ID:
    UniprotID# P51512
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Antigen symbol:
    MMP16
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    Matrix metalloproteinase 16|Membrane-type matrix metalloproteinase 3|MMP-16|Membrane type matrix metalloproteinase 3|Membrane-type-3 matrix metalloproteinase|DKFZp761D112|Membrane type 3 matrix metalloproteinase|Matrix metalloproteinase-16|MMP 16
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    69 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human MMP-16 (amino acids 551-600)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Cat. no.:
    77050-996
  • Packaging:
    Plastic vial

Specifications

About this item

Rabbit polyclonal antibody to MMP-16 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:10000

Type: Primary
Antigen: MMP16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Customers Who Bought This Also Bought