To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:H4F2
- Clonality:Polyclonal
- Gene ID:UniprotID# P62805
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:Histone H4R78me1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Histone H4|HIST1H4|HISTH4H4|H4_HUMAN|HIST2H4|Hist4h4|Histone H4R78me1|H4FN|methyl histone H4
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:11 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Histone H4 (amino acids 13-62)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Histone H4 for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:20000
Type: Primary
Antigen: Histone H4R78me1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat