To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ATLD2
- Clonality:Monoclonal
- Clone:PC10
- Gene ID:UniprotID# P12004
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Drosophila,Chicken,Human,Rat,Mouse
- Antigen symbol:PCNA
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:DNA polymerase delta auxiliary protein|Mutagen-sensitive 209 protein|etID36690.10|HGCN8729|MGC8367|cb16|fa28e03|fb36g03|Cyclin
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant rat PCNA
- Tested applications:IHC-P, IHC-Fr, ICC
- Purification:Protein A chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PC10] antibody to PCNA for Flow Cytometry, IP, WB, IHC-P, IHC-Fr and ICC with samples derived from Human, Mouse, Rat, Chicken and Drosophila.
- Validated Applications: Flow Cytometry, IP, WB, IHC-P, IHC-Fr, ICC
- Recommended Dilutions: Flow Cytometry: 1-4 µg/ml
Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: PC10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat, Chicken, Drosophila