To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:26 kDa cell surface protein TAPA 1
- Clonality:Monoclonal
- Clone:M38
- Gene ID:UniprotID# P60033
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rabbit,Human,Cat
- Antigen symbol:CD81
- Conjugation:Biotin
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CD81 antigen|CD81|26 kDa cell surface protein TAPA1|CD 81|26 kDa cell surface protein TAPA-1|CVID6|CD81_HUMAN|CD81 antigen (target of antiproliferative|CD81 molecule
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage temperature:Store at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:MOLT-4 (human T-ALL cell line)
- Tested applications:IHC-P, ICC
- Purification:This antibody is conjugated with Biotin-LC-NHS under optimum conditions. The reagent is free of unconjugated Biotin.
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [M38] antibody to CD81 (Biotin) for Flow Cytometry, IP, WB, IHC-P and ICC with samples derived from Human, Feline and Rabbit.
- Validated Applications: Flow Cytometry, IP, WB, IHC-P, ICC
- Recommended Dilutions: Flow Cytometry: 1 µg/ml
Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: M38
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Cat, Rabbit