Anti-alpha Tubulin Mouse Monoclonal Antibody [Clone: TU-01] (FITC (Fluorescein Isothiocyanate))
76965-788
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:alpha Tubulin
- Antigen symbol:alpha Tubulin
- Clonality:Monoclonal
- Clone:TU-01
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Pig,Human,Bovine,Dog,Mouse
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q71U36
- Antigen synonyms:H2 ALPHA|Hum a tub2|ALS22|MGC171407|Hum a tub1|bA408E5.3|B ALPHA 1|Alpha-tubulin 1|LIS3
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fraction of tubulin purified from porcine brain by two cycles of polymerization - depolymerization
- Tested applications:ICC
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. no.:76965-788
- Packaging:Plastic vial
Mouse monoclonal [TU-01] antibody to alpha Tubulin (FITC) for Flow Cytometry and ICC with samples derived from Human, Mouse, Bovine, Canine and Porcine.
- Validated Applications: Flow Cytometry, ICC
- Recommended Dilutions: Flow Cytometry: 1-4 µg/ml
Type: Primary
Antigen: alpha Tubulin
Clonality: Monoclonal
Clone: TU-01
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Bovine, Dog, Porcine