Anti-Vimentin Mouse Monoclonal Antibody [Clone: VI-RE/1]
Catalog # 76953-404
Supplier: ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CTRCT30
- Antigen symbol:Vimentin
- Clonality:Monoclonal
- Clone:VI-RE/1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P08670
- Antigen synonyms:Epididymis luminal protein 113|VIME_HUMAN|Vimentin|HEL113|VIM|FLJ36605
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Bacterially expressed full length human Vimentin protein
- Tested applications:ICC, IHC-P
- Purification:Protein A chromatography
- Cat. no.:76953-404
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [VI-RE/1] antibody to Vimentin for Flow Cytometry, WB, ICC, ELISA and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, WB, ICC, ELISA, IHC-P
- Recommended Dilutions: Flow Cytometry: 1-4 µg/ml, WB: 1-2 µg/ml, ICC: 5-10 µg/ml
Type: Primary
Antigen: Vimentin
Clonality: Monoclonal
Clone: VI-RE/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human