To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Alkaline phosphodiesterase I
- Clonality:Monoclonal
- Clone:NP4D6
- Gene ID:UniprotID# O14638
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CD203c
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:100 T
- Form:Liquid
- Antigen synonyms:Ectonucleotide pyrophosphatase/phosphodiesterase 3|dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3)|ARHR2|E NPP 3|B10|E-NPP 3|CD203c|CD203c antigen|dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3)
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:HEK-293 cells transfected with human CD203c
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [NP4D6] antibody to CD203c (APC) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD203c
Clonality: Monoclonal
Clone: NP4D6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human