To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:AOS6
- Clonality:Monoclonal
- Clone:MHD4-46
- Gene ID:UniprotID# Q9NR61
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:DLL4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Delta like protein 4|Delta 4|Delta like 4 homolog|Delta ligand 4|delta ligand 4 precursor|Delta like canonical Notch ligand 4|Delta like 4 protein|delta 4 precursor|Delta like 4
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant soluble human DLL4
- Purification:Protein A chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [MHD4-46] antibody to DLL4 for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1-4 µg/ml
Type: Primary
Antigen: DLL4
Clonality: Monoclonal
Clone: MHD4-46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human