To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:B220
- Clonality:Monoclonal
- Clone:EM-05
- Gene ID:UniprotID# Q8C6Q7
- Host:Rat
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:CD45
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:CD45R|CD45|GP180|Leukocyte common antigen|L-CA|LCA|loc|CD45 antigen|CD 45
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Concentration:0.5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Murine peripheral blood leukocytes
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Packaging:Plastic vial
Specifications
About this item
Rat monoclonal [EM-05] antibody to CD45 (APC) for Flow Cytometry with samples derived from Mouse.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1-2 µg/ml
Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: EM-05
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse