To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Activation B7-1 antigen
- Clonality:Monoclonal
- Clone:16-10A1
- Gene ID:UniprotID# Q00609
- Host:Armenian hamster
- Isotype:IgG2
- Reactivity:Dog,Mouse
- Antigen symbol:CD80
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:B7-1|CD80 antigen|CD28 antigen ligand 1|B7.1|CD28LG|CD28LG1|B lymphocyte activation antigen B7|BB1|CD80
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:CD80-transfected CHO cell line
- Tested applications:IHC-Fr
- Purification:Protein A chromatography
- Packaging:Plastic vial
Specifications
About this item
Armenian hamster monoclonal [16-10A1] antibody to CD80 for Flow Cytometry, IP and IHC-Fr with samples derived from Mouse and Canine.
- Validated Applications: Flow Cytometry, IP, IHC-Fr
- Recommended Dilutions: Flow Cytometry: 1-4 µg/ml
Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: Unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG2
Reactivity: Mouse, Dog