To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:B220
- Clonality:Monoclonal
- Clone:IVA103
- Host:Mouse
- Isotype:IgG1
- Reactivity:Bovine
- Antigen symbol:CD45R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:CD45R|CD45|Glycoprotein|GP180|L CA|Leukocyte common antigen|L-CA|LCA|CD45 antigen
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Bovine leukocytes
- Tested applications:ICC
- Purification:Protein A chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [IVA103] antibody to CD45R for Flow Cytometry, IP and ICC with samples derived from Bovine.
- Validated Applications: Flow Cytometry, IP, ICC
- Recommended Dilutions: Flow Cytometry: 1-4 µg/ml
Type: Primary
Antigen: CD45R
Clonality: Monoclonal
Clone: IVA103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine