To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:AI528660
- Clonality:Monoclonal
- Clone:MEM-226
- Gene ID:UniprotID# P17813
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Rat
- Antigen symbol:CD105
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 T
- Form:Liquid
- Antigen synonyms:CD 105|Eng|Endoglin|AI662476|EGLN_HUMAN|CD105 antigen|FLJ41744|CD105|END
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant Vaccinia virus containing the human CD105 cDNA
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [MEM-226] antibody to CD105 (PE) for Flow Cytometry with samples derived from Human and Rat.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD105
Clonality: Monoclonal
Clone: MEM-226
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat