To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Acylation stimulating protein cleavage product
- Clonality:Monoclonal
- Clone:C3D/2891
- Gene ID:UniprotID# P01024
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Antigen symbol:C3d
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:AHUS5|Complement C3c alpha' chain fragment 2|ARMD9|C3|Complement C3|ASP|C3d|C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1|CO3_HUMAN
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:35 kDa (predicted)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human Complement C3 protein
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [C3D/2891] antibody to C3d for ELISA with samples derived from Human.
- Validated Applications: ELISA
Type: Primary
Antigen: C3d
Clonality: Monoclonal
Clone: C3D/2891
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human