To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:campomelic dysplasia autosomal sex reversal
- Clonality:Recombinant
- Clone:SOX9/2287R
- Gene ID:UniprotID# P48436
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:SOX9
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:CMD 1|CMD1|CMPD 1|SOX 9|SOX9_HUMAN|SRA1|SOX9|CMPD1|SRA 1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:56 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human SOX9 protein
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [SOX9/2287R] antibody to SOX9 for ELISA with samples derived from Human.
- Validated Applications: ELISA
- Recommended Dilutions: ELISA: 1 to 5 µg/ml
Type: Primary
Antigen: SOX9
Clonality: Recombinant
Clone: SOX9/2287R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human