To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:B220
- Clonality:Monoclonal
- Clone:111-1C5
- Gene ID:UniprotID# P08575
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD45RA
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CD45R|CD45|CD45RA|GP180|Leukocyte common antigen|L-CA|LCA|CD45 antigen|EC 3.1.3.48
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:205 - 220 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Stimulated human leukocytes
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [111-1C5] antibody to CD45RA for Flow Cytometry, IF, WB and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: 111-1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human