To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Acid phosphatase 5 tartrate resistant
- Clonality:Monoclonal
- Clone:SPM601
- Gene ID:UniprotID# P13686
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Rat,Mouse
- Antigen symbol:Tartrate Resistant Acid Phosphatase
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:SPENCDI|Acid phosphatase 5, tartrate resistant|ACP5|T5ap|serum band 5 tartrate-resistant acid phosphatase|Tartrate resistant acid ATPase|EC 3.1.3.2|MGC117378|PPA5_HUMAN
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:35 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human ACP5 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [SPM601] antibody to Tartrate Resistant Acid Phosphatase for Flow Cytometry, IF and IHC-P with samples derived from Human, Mouse and Rat.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Tartrate Resistant Acid Phosphatase
Clonality: Monoclonal
Clone: SPM601
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse, Rat