To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:HPV16/1295 + HPV18/1297
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:HPV-16
- Antigen symbol:HPV-16 E6 + HPV-18 E6
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Form:Liquid
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:16 kDa/17 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:HPV18 E6-beta Galactosidase fusion protein
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [HPV16/1295 + HPV18/1297] antibody to HPV-16 E6 + HPV-18 E6 for ELISA with samples derived from HPV-16.
- Validated Applications: ELISA
Type: Primary
Antigen: HPV-16 E6 + HPV-18 E6
Clonality: Monoclonal
Clone: HPV16/1295 + HPV18/1297
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: HPV-16