To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:26 kDa cell surface protein TAPA 1
- Clonality:Recombinant
- Clone:r1.3.3.22
- Gene ID:UniprotID# P60033
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TAPA1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CD81 antigen|CD81|26 kDa cell surface protein TAPA1|CD 81|26 kDa cell surface protein TAPA-1|CVID6|CD81_HUMAN|CD81 antigen (target of antiproliferative|CD81 molecule
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:26 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human CD81 protein
- Tested applications:Functional Studies, IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [r1.3.3.22] antibody to TAPA1 for Functional Studies, Flow Cytometry, IF, WB and IHC-P with samples derived from Human, Mouse and Rat.
- Validated Applications: Functional Studies, Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 0.5 to 1 µg/million cells, IF: 0.5 to 1 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 2 to 4 µg/ml
Type: Primary
Antigen: TAPA1
Clonality: Recombinant
Clone: r1.3.3.22
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat