To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:B2M
- Clonality:Monoclonal
- Clone:246-E9.E7
- Gene ID:UniprotID# P61769
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Antigen symbol:beta 2 Microglobulin
- Conjugation:Unconjugated
- Size:100 µg
- Form:Liquid
- Antigen synonyms:CDABP0092|Beta 2 microglobulin precursor |Hdcma22p|Beta chain of mhc class 1 proteins |Beta chain of MHC class I molecules|beta 2 Microglobulin|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin|B2MG_HUMAN
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:12 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Human PBL s from a T-cell acute lymphoblastic leukemia (T-ALL) patient
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [246-E9.E7] antibody to beta 2 Microglobulin with samples derived from Human.
Type: Primary
Antigen: beta 2 Microglobulin
Clonality: Monoclonal
Clone: 246-E9.E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human