To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cell proliferation inducing gene 46 protein
- Clonality:Recombinant
- Clone:KRT18/2819R
- Gene ID:UniprotID# P05783
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse,Cat,Marmoset,Hamster,Pig,Horse,Sheep,Goat,Ferret,Human,Bovine,Dog,Rat
- Antigen symbol:Cytokeratin 18
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|Cytokeratin endo B|Cell proliferation-inducing gene 46 protein|CK 18|CK-18|CYK 18
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:45 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human KRT18 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [KRT18/2819R] antibody to Cytokeratin 18 for Flow Cytometry, WB, IF and IHC-P with samples derived from Human, Mouse, Rat, Hamster, Ferret, Bovine, Porcine, Feline, Canine, Sheep, Goat, Horse and Marmoset Monkey.
- Validated Applications: Flow Cytometry, WB, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 18
Clonality: Recombinant
Clone: KRT18/2819R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Ferret, Bovine, Porcine, Cat, Dog, Sheep, Goat, Horse, Marmoset Monkey