To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Akp2
- Clonality:Monoclonal
- Clone:PL8-F6
- Gene ID:UniprotID# P05187
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Antigen symbol:Placental Alkaline Phosphatase
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Alkaline phosphatase placental type|Alkaline phosphatase liver/bone/kidney isozyme|Alkaline phosphatase intestinal precursor|Alkaline phosphatase placental like|Alkaline Phosphatase|Alkaline phosphatase placental|Alkaline phosphatase liver/bone/kidney|Alkaline phosphatase intestinal|Alkaline phosphatase placental like 2
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:70 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Purified human PLAP protein
- Tested applications:IHC
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PL8-F6] antibody to Placental Alkaline Phosphatase for WB, Flow Cytometry, IF and IHC with samples derived from Human.
- Validated Applications: WB, Flow Cytometry, IF, IHC
- Recommended Dilutions: WB: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml, IHC to Fr: 1 to 2 µg/ml
Type: Primary
Antigen: Placental Alkaline Phosphatase
Clonality: Monoclonal
Clone: PL8-F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human