To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:2 phospho D glycerate hydrolyase
- Clonality:Monoclonal
- Clone:ENO2/1375
- Gene ID:UniprotID# P09104
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Rat,Mouse
- Antigen symbol:Neuron Specific Enolase
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Enolase 2|Enolase 2 (gamma, neuronal)|ENO2|Enolase 2 gamma neuronal|ENOG|ENOG_HUMAN|Enolase2|Eno 2|2-phospho-D-glycerate hydro-lyase
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:~50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide, within amino acids 416-433, of human NSE gamma. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:76951-932
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [ENO2/1375] antibody to Neuron Specific Enolase for WB and IHC-P with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 0.2 to 0.4 µg/ml
Type: Primary
Antigen: Neuron Specific Enolase
Clonality: Monoclonal
Clone: ENO2/1375
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse, Rat